Skip to Content
Merck
All Photos(1)

Key Documents

392170

Sigma-Aldrich

Methyl 3,4-O-isopropylidene-L-threonate

97%

Synonym(s):

R,4S)-2,2-Dimethyl-α-hydroxy-1,3-dioxolane-4-acetic acid methyl ester, 3,4-O-Isopropylidene-L-threonic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O5
CAS Number:
Molecular Weight:
190.19
Beilstein:
7862
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

optical activity

[α]20/D +18.5°, c = 2 in acetone

refractive index

n20/D 1.446 (lit.)

bp

125 °C/12 mmHg (lit.)

density

1.175 g/mL at 25 °C (lit.)

SMILES string

COC(=O)[C@H](O)[C@@H]1COC(C)(C)O1

InChI

1S/C8H14O5/c1-8(2)12-4-5(13-8)6(9)7(10)11-3/h5-6,9H,4H2,1-3H3/t5-,6+/m0/s1

InChI key

JSFASCPVVJBLRD-NTSWFWBYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Used to prepare a dihydrofuran substrate in the total synthesis of (-)-echinosporin.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 114, 2567-2567 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service