Skip to Content
Merck
All Photos(1)

Key Documents

288322

Sigma-Aldrich

Methyl 3,3-dimethoxypropionate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2CHCH2CO2CH3
CAS Number:
Molecular Weight:
148.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.41 (lit.)

bp

77 °C/20 mmHg (lit.)

density

1.045 g/mL at 25 °C (lit.)

functional group

acetal
ester
ether

SMILES string

COC(CC(=O)OC)OC

InChI

1S/C6H12O4/c1-8-5(7)4-6(9-2)10-3/h6H,4H2,1-3H3

InChI key

SMCVPMKCDDNUCQ-UHFFFAOYSA-N

General description

Methyl 3,3-dimethoxypropionate was used in the synthesis of 3-indolyl α,β-unsaturated carbonyl compounds[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A practical synthesis of 3-indolyl α, β-unsaturated carbonyl compounds.
Wang W and Ikemoto T.
Tetrahedron Letters, 46(22), 3875-3878 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service