- Cytotoxic effects of protease inhibitors on human cells. 2. Effect of elastatinal.
Cytotoxic effects of protease inhibitors on human cells. 2. Effect of elastatinal.
Cancer letters (1980-10-01)
T Yagi, K Ishizaki, H Takebe
PMID7191773
RESUMEN
Effects of elastatinal, a protease inhibitor, on DNA repair and its cytotoxicity were investigated in normal human, xeroderma pigmentosum and Chinese hamster V79 cells. There were no effects of elastatinal on DNA repair and UV-induced sister chromatid exchanges, and all 3 cell lines demonstrated essentially the same sensitivity to the cytotoxic effect of elastatinal. These results were quite different from those by antipain, another protease inhibitor, and caffeine.
MATERIALES