Merck
  • [Synthesis and antibacterial and antitumoral activity of some methylhydrazonium salts of pyrimidine bases and their analogues (author's transl)].
[Synthesis and antibacterial and antitumoral activity of some methylhydrazonium salts of pyrimidine bases and their analogues (author's transl)].
Die Pharmazie (1982-05-01)
E Golovinsky, G Karamanov, N Spassovska, L Maneva, A Spassov
RESUMEN
On reacting pyrimidine metabolites containing a carboxyl or sulfhydryl group with methylhydrazine or N-benzyl-N'-methylhydrazine, various methylhydrazonium salts of these metabolites were synthetized, e.g., the salts of 5-fluororotic acid, 5-azaorotic acid, 2-thiouracil-5-carboxylic acid and 2-thio-6-azathymine. Some of these salts exhibited a more marked antibacterial activity against St. aureus and several of its mutants as well as a greater antitumoral activity against different transplantable tumours than their single components.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
5-Carboxy-2-thiouracil