Documentos clave
509639
Syringes Fitted with Luer Lock Valve
volume 25 mL, 25MDR-VLLMA-GT, needle size (not included)
About This Item
Código UNSPSC:
41122001
Materiales
glass syringe
tamaño de la aguja
(not included)
Características
Pre-fitted with Luer Lock Valve
envase
pkg of 1 ea
fabricante / nombre comercial
SGE 009472
volumen
25 mL
Descripción general
<UL><LI>SGE Syringes Fitted with Luer Lock Valve [volume 25 mL, needle size (not included)], Model: 25MDR-VLLMA-GT.</LI><LI>Composed of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.</LI><LI>Autoclavable syringes but the plunger removal required.</LI><LI>Feature an interference fit between the plunger tip and the barrel that enhances the gas sealing ability of the syringe.
  • Maximum inertness for liquid or gas samples.
  • Suitable for manual dispenser/diluter applications.
  • High accuracy of dispensed volume.
  • A wide variety of terminations.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)