Documentos clave
Z329282
BRAND® magnetic stirring bar, PTFE, oval
L 50 mm, diam. 20 mm
About This Item
UNSPSC Code:
41103806
eCl@ss:
32100100
NACRES:
NB.72
material
PTFE-coated
oval
manufacturer/tradename
BRAND 137315
L
50 mm
diam.
20 mm
General description
Ideal for vessels with round bottom, e.g. round-bottom flasks.
Other Notes
BR137315 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG