Documentos
HS120177
Sharps container
1 lb, red polypropylene
About This Item
UNSPSC Code:
41123300
NACRES:
NB.51
material
red polypropylene
feature
Holds sharps
packaging
pack of 18 ea
manufacturer/tradename
Heathrow Scientific HS120177
size
115x115x192 mm
capacity
1 lb
General description
The Sharps Container is easy to use and transport. Provides safe disposal of syringes, needles and other small sharps.
  • Stackable to reduce storage space before use
  • 1.0 L capacity (Quart) with a clear lid to view fill level
  • Slide access lid for temporary and final closures for safety and security
  • Built-in needle removal port, preventing direct contact with sharps
  • Compatible with the Droplet Blood Collection Tray
  • Measures 4.5 x 4.5 x 7.5 in (11.5 x 11.5 x 19.2 cm)
  • Pack of 18
For use with
Referencia del producto