EP951033707
Eppendorf® Deepwell Plate 96/2000 uL
sterile, volume 2000 μL, green border with clear wells, pkg of 80 plates (10 bag x 8 plates)
About This Item
Productos recomendados
material
colorless wells
round bottom wells
sterility
sterile
feature
green border with clear wells
packaging
pkg of 80 plates (10 bag x 8 plates)
manufacturer/tradename
Eppendorf® 951033707 (North America)
technique(s)
PCR: suitable
size
96 well
volume
2000 μL
working volume
2 mL
Categorías relacionadas
Legal Information
Eppendorf is a registered trademark of Eppendorf AG