Key Documents
EP5895180000
Rotor for Eppendorf® 5910 R Centrifuge
S-4x400, incl. 4 x 400 mL round buckets
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 0.2 mL to 400 mL tubes and bottles
S-4x400, incl. 4 x 400 mL round buckets
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5895180000
parameter
51,000 max. RCF (5,263 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5910R
General description
Rotor for Eppendorf(R)5910R Centrifuge
Legal Information
Eppendorf is a registered trademark of Eppendorf AG