Documentos clave
EP0030140516
Storage Box for Eppendorf® Tubes
size 9 × 9, 81 screw cap (cryo.) tubes 1-2 mL
About This Item
Código UNSPSC:
41123300
NACRES:
NB.12
Materiales
polypropylene
Características
81 screw cap (cryo.) tubes 1-2 mL
Holds 81 x 1-2 mL tubes
fabricante / nombre comercial
Eppendorf® 0030140516 (Global)
Altura
53 mm
tamaño
9 × 9
Descripción general
The comprehensive modular concept allows selection of the optimal freezer box combinations for individual storage demands. The outer footprint dimensions (133 mm) of these freezer boxes are compatible with common freezer rack systems and the different inner grid variants provide for an optimal fit to all typical tube formats: cryogenic tubes, microcentrifuge tubes, 15, 25 and 50 mL conical tubes and other laboratory vessels. This space-saving approach together with the application benefits resulting from the listed product features help to optimize your longterm storage and archiving of samples.
Información legal
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)