Documentos clave
SK-N-BE(2)
95011815, human nerve, Epitheilioid & neuroblastoid
About This Item
UNSPSC Code:
41106514
product name
SK-N-BE(2), 95011815
biological source
human nerve
growth mode
Adherent
karyotype
Modal no. 44
morphology
Epitheilioid & neuroblastoid
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
metastasis
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian neuroblastoma
Cell Line Description
Established from a bone marrow biopsy of a 22-month-old male with disseminated neuroblastoma in 1972. The morphology varies with sub-population from cells with long processes through small, flattened, epithelioid cells. Dense cultures tend to form loosely adherent aggregates. They are multipotential with regard to neuronal enzyme expression and display a high capacity to convert tyrosine to dopamine.
Application
Chemotherapy development, chromosome studies, drug resistance, differentiation studies.
Culture Medium
EMEM (EBSS) + 1% Non Essential Amino Acids (NEAA) : Ham′s F12 (1:1) + 2mM Glutamine + 15% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:10 to 1:25 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.