Key Documents
Z414190
Sigma-Aldrich
Aldrich® pour-out adapter
male joint: ST/NS 29/32
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
male joint: ST/NS29/32
General description
For pouring from flasks or funnels without pouring over ground joint.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC