Documentos
Z247243
Coors porcelain crucible cover
cap size E, I.D. 74 mm
About This Item
UNSPSC Code:
41122200
NACRES:
NB.43
cap size
E
packaging
pkg of 10 ea
manufacturer/tradename
Coorstek 66126
I.D.
74 mm
General description
Coors porcelain crucible cover is glazed all over except for the rim.
Features and Benefits
  • Ensures great resistance to thermal-shock
  • Durable
  • Rigid
  • Sustained refractory qualities
Other Notes
Covers may not fit exactly on crucibles due to unavoidable wall thickness variations within the crucible.
Legal Information
Coors is a trademark of Coorstek, Inc.