Documentos clave
Z223352
Sigma-Aldrich
Aldrich® in-line oil bubbler
joint: ST/NS 24/40
Sinónimos:
bubblers, in-line bubblers, scientific bubblers
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
joint
joint: ST/NS24/40
capacity
10 mL
Categorías relacionadas
General description
For precise N2 pressure control during inert atmosphere reactions. Connect reaction vessel to in-line joint or use with a ballast bulb to keep pressure constant.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)