Yes, this rack will hold 40 tubes having a diameter of 13 -16 mm.
Z176974
Scienceware® serum-vial support
Holds 40 x 13-16 mm tubes
About This Item
Productos recomendados
Materiales
polypropylene
Características
Holds 40 x 13-16 mm tubes
envase
pack of 1 ea
fabricante / nombre comercial
Bel-Art F18513-1640
L × An × Al
10 in. × 4 3/8 in. × 3 1/2 in.
Información legal
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
-
It says it holds 13-16mm tubes - does this refer to the diameter of the tubes?
1 answer-