Documentos clave
Z176664
Sigma-Aldrich
Aldrich® cold plate
AC/DC input 115 V AC, Analog cold plate
About This Item
Código UNSPSC:
41121800
esterilidad
non-sterile
disponibilidad
not available in EU
color
none
Descripción general
These portable, low-temperature control devices utilize efficient, solid-state, thermoelectric modules to control temperatures from +25 to -20 °C. Unique, dual heat exchanger utilizes either fan forced air or tap water for cooling. Connects to water source using 1/4 in. I.D. tubing. Stirring models have a built-in, adjustable-speed, magnetic stirrer to keep solutions at equilibrium. Useful for reaction kinetics, recrystallizations, cold traps, freezing point determinations, controlling exothermic reactions, condenser for distillations, freezing biological and blood specimens, cold bath or storage chambers. Not CE compliant.
Specifications:
  • Rapid cooling to -10 °C (forced air) or -20 °C (tap water), 130 BTU/h initial rate
  • Cooling to -40 °C possible by circulating coolant
  • Equilibrium reached in 2 to 3 min
  • 3 in. diam. aluminum top plate handles solutions up to 200 mL
  • 18 gauge aluminum cabinet, 7 W × 5 D × 5 in. H
  • Wt = 8 lb
Analog model has adjustable power supply to control cooling.
Otras notas
The use of insulated, flat-bottom glassware and aluminum cooling blocks maximizes heat transfer rates from media to cold plate and are highly recommended for use with this equipment.
Información legal
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Matthew W Hale et al.
Behavioural brain research, 359, 428-439 (2018-11-24)
Evidence suggests that affective disorders are associated with altered thermoregulation, and it has been hypothesized that therapeutic strategies targeting body-to-brain thermosensory systems may be effective for treating depression. Consistent with this hypothesis, a recent randomized, double blind, placebo-controlled clinical trial