Documentos
Z125350
Ace-Thred Thermometer/Tubing Adapter
Straight, joint: ST/NS 10/18, thread I.D. 6.5 mm
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
label
A
joint
joint: ST/NS10/18
manufacturer/tradename
Ace Glass 502824
thread I.D.
6.5 mm
General description
Threaded nylon bushing with FETFE® O-ring.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
replacement part
Referencia del producto
Descripción
Precios
Visite la Librería de documentos
Los clientes también vieron
Slide 1 of 1