WHA10418506
Whatman® Nuclepore™ Track-Etched Membranes
polycarbonate filter, filter diam. 25 mm, pore size 12 μm, non-sterile, case of 100 ea
About This Item
UNSPSC Code:
41104921
NACRES:
NB.24
Productos recomendados
material
polycarbonate filter
sterility
non-sterile
packaging
case of 100 ea
manufacturer/tradename
Cytiva 10418506
filter diam.
25 mm
pore size
12 μm
Application
- Epifluorescence microscopy
- Environmental analysis
- Cell biologyEPA testing
- Fuel testing
- Bioassays
- Parasitology
- Air analysis
- Water microbiology
Legal Information
Nuclepore is a trademark of Cytiva
Whatman is a registered trademark of Cytiva