SYNF309500
Synthware™ single-neck round-bottom flask
500 mL, joint: ST/NS 19/22
Sinónimos:
Synthware F309500, glass round flask
About This Item
Productos recomendados
material
borosilicate glass flask
round bottom
single-neck
joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F309500
capacity
500 mL
500 mL
Categorías relacionadas
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)