Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.24
UNSPSC Code:
41116107
grade
pharmaceutical primary standard
agency
USP
API family
etodolac
form
solid
manufacturer/tradename
USP
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Etodolac Peak Identification Mixture USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Other Notes
- Etodolac [CAS 41340-25-4]
- 1-Methyl Etodolac analog [CAS 109518-50-5]
- 8-Methyl Etodolac analog [CAS 41340-19-6]
- 8-Isopropyl Etodolac analog [CAS 57917-63-2]
- 8-Propyl Etodolac analog [CAS 57817-27-3]
- 7-Ethyltrypthanol [CAS 41340-36-7]
- 8-Desethyl Etodolac [CAS 41339-67-7]
- 1-Isopropyl Etodolac analog [CAS 849630-65-5]
- 1-Propyl Etodolac analog [CAS 57816-83-8]
- Etodolac dimer
- Decarboxyetodolac [CAS 115066-03-0]
- Etodolac Methyl ester [CAS 122188-02-7]
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
