Key Documents
21015
Supelco
Female Luer Coupler
About This Item
UNSPSC Code:
12190000
Pricing and availability is not currently available.
packaging
pkg of 20 ea
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
compatibility
for use with male luer to male luer
General description
Female Luer Coupler is an accessory for use with LpDNPH Cartridges suitable for air monitoring applications.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Are these female Luer couplers sterile packed?
    1 answer
    1. The female Luer couplers in Product No. 21015 are not sterile-packaged.
      Helpful?