ZRB1973
Anti-Ubiquityl H2B-Lys120 Antibody, clone 1F17 ZooMAb® Rabbit Monoclonal
recombinant, expressed in HEK 293 cells
Synonym(s):
Histone H2B type 1-B;H2B-clustered histone 3;Histone H2B.1;Histone H2B.f; H2B/f
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1F17, recombinant monoclonal
description
1F17 Clone
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 13.95 kDa
observed mol wt ~24 kDa
purified by
using Protein A
species reactivity
rat, human
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
immunohistochemistry: suitable
inhibition assay: suitable
western blot: suitable
isotype
IgG
epitope sequence
C-terminal
Protein ID accession no.
UniProt accession no.
greener alternative category
, Aligned
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
ubiquitination (Lys120)
Gene Information
human ... H2BC3(3018)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in HeLa cell acid extract.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Ubiquityl Histone H2B-Lys120 in HeLa cell acid extract.
Tested Applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Ubiquityl Histone H2B-Lys120 in HL-60 cell and Rat2 cell acid extracts .
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Ubiquityl Histone H2B-Lys120 in human liver tissue sections.
Peptide Inhibition Assay: Target band detection in a lysate from HeLa cell acid extract was prevented by pre-blocking of a representative lot with Ubiquityl Histone H2B-Lys120 peptide, but not the corresponding non- Ubiquityl H2B peptide.
Affinity Binding Assay: A representative lot of this antibody bound Ubiquityl Histone H2B-Lys120 peptide with a KD of 3.5 x 10-8 in an affinity binding assay.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service