Key Documents
Z744319
Wilmad® 150 ml Suprasil® Large Volume Liquid Nitrogen Dewar
offset for Bruker®, pack of 1 ea
About This Item
UNSPSC Code:
41121804
NACRES:
NB.15
packaging
pack of 1 ea
manufacturer/tradename
Wilmad WG-853-B-Q
capacity
150 mL
General description
Dewar Flasks provide a convenient alternative to Variable Temperature systems for studies at 77 K, and are silvered around the reservoir.
Features and Benefits
150 mL flask lasts 30 minutes after a filling
Legal Information
Bruker is a registered trademark of Bruker-Physik AG
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries