Key Documents
Z743579
Speedy-Foil® aluminium squares interleaved with paper
L × W 2 in. × 2 in., pack of 500 sheets
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
Pricing and availability is not currently available.
packaging
pack of 500 sheets
manufacturer/tradename
Diversified Biotech FOIL-1000
L × W
2 in. × 2 in.
General description
PRE-CUT FOIL AT YOUR FINGER TIPS
  • Faster, easier and aseptic: grab and cover
  • Pre-cut to fit common laboratory containers
  • No scissors and less waste
  • 1 mil thick for a sharp, crisp dead-fold providing a tighter fit
  • Each square is interleaved for physical separation
  • Meets FDA requirements for direct contact with pharmaceuticals; RNase and DNase free
Legal Information
Speedy-Foil is a registered trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. What is the purity of the foil? 
    1 answer
    1. This item is a product of Diversified Biotech. According to the manufacturer, these aluminum pre-cut squares are essentially 100% aluminum.
      Helpful?