Z743373
Direct Thermal Cryo-Tags®
Cryo-Spot, white, diam. Type="regular">1/2 in., pkg of 4000 roll of
Synonym(s):
cryogenic vial labels, freezer labels
About This Item
Recommended Products
packaging
pkg of 4000 roll of
manufacturer/tradename
Diversified Biotech DTCR-4500
parameter
temp. range at 70 to -196 °C
technique(s)
cryopreservation: suitable
diam.
Type="regular">1/2 in.
color
white
compatibility
Important note for Customers outside of USA: Our Direct Thermal labels are designed to work with U.S. Model Dymo printers only. Labels are not compatible with European DYMO printers.
Looking for similar products? Visit Product Comparison Guide
General description
Tough - Labels adhere to most plastics, glass and metals without cracking, peeling or degrading. Also resistant to scuffing and smearing
Compatible - Tubes labeled with Direct Thermal Cryo-Tags easily slide in and out of centrifuge rotors without binding
Specially Designed - Labels accept printing from direct thermal printers such as GoDEX DT200 Direct printers. Diversified Biotech recommends the Label LIVE app for printing in conjunction with the GoDEX DT200 printer.
Barcoding - Print popular barcode formats for fast and easy sample identification
Economical - Limit label waste by printing only the quantity required. Available in White, Blue, Green and Yellow
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service