Skip to Content
Merck
All Photos(1)

Key Documents

SML3708

Sigma-Aldrich

L-Meta-tyrosine

≥98% (HPLC)

Synonym(s):

(S)-2-Amino-3-(3-hydroxyphenyl)propanoic acid, 3-Hydroxy-L-phenylalanine, L-3-Hydroxyphenylalanine, L-m-Tyrosine, m-Tyr, meta-Tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO3
CAS Number:
Molecular Weight:
181.19
MDL number:
UNSPSC Code:
12352209
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

2 mg/mL, clear (0.1N HCl)

storage temp.

-10 to -25°C

SMILES string

[N+H3][C@@H](Cc1cc(ccc1)O)C(=O)[O-]

InChI

1S/C9H11NO3/c10-8(9(12)13)5-6-2-1-3-7(11)4-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1

Biochem/physiol Actions

L-Meta-tyrosine is metabolite of L-Phenylalanine generated within the living organism by hydroxyl free radical. It is a marker of oxidative stress. Circulating L-Meta-tyrosine cold be incorporated into protein through the binding to the tRNAPhe. In plants L-Meta-tyrosine (m-Tyr) is also synthesized enzymatically and has been shown to act as a natural herbicide. It inhibits the growth of plants. L-Meta-tyrosine is a potent and selective competitive inhibitor of Enterobacteriaceae (Citrobacter, Proteus, Erwinia) tyrosine phenol-lyase that exhibits a renoprotective effect in diabetic mice. L-Meta-tyrosine decreases plasma phenyl sulfate and indoxyl sulfate levels and reduces albuminuria in diabetic mice models. Also, L-Meta-tyrosine inhibits bacterial tryptophan indole-lyase. It does not alter composition of microbiota in renal failure mice model.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcin Tyminski et al.
Plants (Basel, Switzerland), 10(12) (2021-12-29)
L-Tyrosine (Tyr) is one of the twenty proteinogenic amino acids and also acts as a precursor for secondary metabolites. Tyr is prone to modifications, especially under conditions of cellular redox imbalance. The oxidation of Tyr precursor phenylalanine leads to the
Koichi Kikuchi et al.
Nature communications, 10(1), 1835-1835 (2019-04-25)
Diabetic kidney disease is a major cause of renal failure that urgently necessitates a breakthrough in disease management. Here we show using untargeted metabolomics that levels of phenyl sulfate, a gut microbiota-derived metabolite, increase with the progression of diabetes in
Judit Mohás-Cseh et al.
Biomedicines, 10(5) (2022-05-29)
A link between oxidative stress and insulin resistance has been suggested. Hydroxyl free radicals are known to be able to convert phenylalanine (Phe) into the non-physiological tyrosine isoforms ortho- and meta-tyrosine (o-Tyr, m-Tyr). The aim of our study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service