Skip to Content
Merck
All Photos(1)

Key Documents

SML3622

Sigma-Aldrich

CAY10444

≥98% (HPLC)

Synonym(s):

(4R)-2-Undecyl-4-thiazolidinecarboxylic acid, 2-Undecylthiazolidine-4(R)-carboxylic acid, BML 241, BML-241, BML241, CAY 10444, CAY-10444

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H29NO2S
CAS Number:
Molecular Weight:
287.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

CAY10444 is a selective sphingosine-1-phosphate (S1P) receptor 3 (S1P3; EDG3) antagonist that inhibits 1 µM S1P-induced calcium mobilization in HeLa transfectants that express S1P3 over those expressing S1P1/CD363/EDG1 (37.6% vs 6.92% inhibition by 10 µM CAY10444).
Selective sphingosine-1-phosphate (S1P) receptor 3 (S1P3; EDG3) antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuuki Koide et al.
Journal of medicinal chemistry, 45(21), 4629-4638 (2002-10-04)
Sphingosine-1-phosphate (S1P) is an intracellular second messenger and an extracellular mediator through endothelial differentiation gene (EDG) receptors, which are a novel class of G-protein-coupled receptors. Although EDG has attracted much attention because of its various roles, no selective agonists or
Alessandra Bigi et al.
The FEBS journal, 290(1), 112-133 (2022-07-20)
Soluble oligomers arising from the aggregation of the amyloid beta peptide (Aβ) have been identified as the main pathogenic agents in Alzheimer's disease (AD). Prefibrillar oligomers of the 42-residue form of Aβ (Aβ42 O) show membrane-binding capacity and trigger the
Inhibiting Sphingosine 1-Phosphate Receptor Subtype 3 Attenuates Brain Damage During Ischemia-Reperfusion Injury by Regulating nNOS/NO and Oxidative Stress
Frontiers in Neuroscience, 16, 838621-838621 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service