Skip to Content
Merck
All Photos(1)

Key Documents

SML2967

Sigma-Aldrich

Gp91ds-tat, scrambled trifluoroacetate

≥95% (HPLC)

Synonym(s):

Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-Cys-Leu-Arg-Ile-Thr-Arg-Gln-Ser-Arg-NH2 trifluoroacetate, Gp91 ds-tat, scrambled trifluoroacetate, NADPH oxidase 2 docking sequence-tat trifluoroacetate, NOX2 ds-tat, scrambled trifluoroacetate, NOX2ds-tat, scrambled trifluoroacetate, RKKRRQRRR-CLRITRQSR-NH2 trifluoroacetate, scrambled Gp91phox docking sequence-tat trifluoroacetate, scrambled NOX2 docking sequence-tat trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C98H190N50O22S · xC2HF3O2
Molecular Weight:
2452.94 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

film or powder

color

white to off-white

storage temp.

−20°C

Biochem/physiol Actions

Inactive control peptide for the NADPH oxidase inhibitor peptide gp91ds-tat (NOX2ds-tat). The control peptide is composed of a membrane-permeant HIV-1 Tat protein transduction domain sequence (aa49-57) N-terminal to a scrambled sequence of murine gp91phox (NOX2) aa86-94.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F E Rey et al.
Circulation research, 89(5), 408-414 (2001-09-05)
We previously reported enhanced expression of the p67(phox) and gp91(phox) components of NAD(P)H oxidase in angiotensin (Ang) II-induced hypertension, suggesting de novo assembly in response to Ang II. To examine the direct involvement of NAD(P)H oxidases in Ang II-induced O(2)(-)
Dina Vara et al.
Haematologica, 104(9), 1879-1891 (2019-01-27)
The regulation of platelets by oxidants is critical for vascular health and may explain thrombotic complications in diseases such as diabetes and dementia, but remains poorly understood. Here, we describe a novel technique combining electron paramagnetic resonance spectroscopy and turbidimetry
Boris Sabirzhanov et al.
Brain, behavior, and immunity, 80, 73-87 (2019-02-27)
NADPH oxidase (NOX2) is an enzyme that induces reactive oxygen species (ROS) and serves as a switch between the pro-inflammatory and neurorestorative microglial/macrophage phenotypes; such changes play an important role in neuropathic pain and motor dysfunction. Increased NOX2 expression after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service