Skip to Content
Merck
All Photos(1)

Key Documents

SAB4503698

Sigma-Aldrich

Anti-CD97 β (Cleaved-Ser531) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CD97, TM7LN1, heterodimeric receptor associated with inflammation, leukocyte antigen CD97, seven-transmembrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 36 kDa
antigen 92 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

proteolytically cleaved (Ser531)

Gene Information

human ... CD97(976)

Related Categories

General description

Anti-CD97 β (Cleaved-Ser531) Antibody detects endogenous levels of fragment of activated CD97 β (Cleaved-Ser531) protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human CD97 beta.

Immunogen Range: 512-561

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service