Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500500

Sigma-Aldrich

Anti-CSFR antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CD115, CSF-1-R, CSF1R, CSFMR, c-fms

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 107 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CSF1R(1436)

General description

Colony stimulating factor 1 receptor (CSF-1R) is encoded by the c-fms proto-oncogene mapped to human chromosome 5q33. The encoded protein acts as a receptor for CSF1 expressed in non-hematopoietic cells, such as macrophages, epithelial and fibroblasts cells and tumor cells.

Immunogen

The antiserum was produced against synthesized peptide derived from human CSFR.

Immunogen Range: 531-580

Biochem/physiol Actions

Colony stimulating factor 1 receptor (CSF-1R)/ CSF-1 ligand system plays a vital role in the regulation of macrophage physiology in immune system.The encoded protein functions as a receptor and mediates the function of its ligand, colony stimulating factor 1, a cytokine involved in regulation of production, differentiation, and function of macrophages. Experimental analysis revealed that CSF-1/CSF-1R is overexpressed in patients with nasopharyngeal carcinoma when compared to patients with nasopharyngitis. Activated CSF-1R, is essential for monocyte differentiation to macrophages by oxidized low-density lipoprotein (Ox-LDL), which is a vital step in early atherogenesis. This protein is also associated with regulation of hematopoiesis. Deletion or mutation of this gene might be involved in the pathogenesis of myelodysplastic syndromes or of acute nonlymphocytic leukemia (ANLL).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ox-LDL induces monocyte-to-macrophage differentiation in vivo: Possible role for the macrophage colony stimulating factor receptor (M-CSF-R).
Fuhrman B, et al.
Atherosclerosis, 196(2), 598-607 (2008)
Evidence for the involvement of GM-CSF and FMS in the deletion (5q) in myeloid disorders.
Le Beau MM, et al.
Science, 231(4741), 984-987 (1986)
Overexpression of CSF-1R in nasopharyngeal carcinoma.
Huang L, et al.
Romanian Journal of Morphology and Embryology, 56(4), 1279-1283 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service