Skip to Content
Merck
All Photos(1)

Documents

SAB3700788

Sigma-Aldrich

Anti-Ferret IgA (α-chain specific)-Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

ALP, Alk Phos

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

ferret

concentration

1.0 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Ferret IgA coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Alkaline Phosphatase (calf intestine), Anti-Goat Serum, Ferret IgA and Ferret Serum. Specificity was confirmed by ELISA at less than 1% cross reactivity against other Ferret heavy or light chain isotypes.

Immunogen

Ferret IgA alpha heavy chain

Physical properties

Antibody format: IgG

Physical form

Supplied in 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0 with 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service