Skip to Content
Merck
All Photos(1)

Documents

M051000

Sigma-Aldrich

DMT-dU Phosphoramidite

Synonym(s):

DMT-dU amidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H47N4O8P
Molecular Weight:
730.79
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

product line

Proligo Reagents

Quality Level

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

λ

conforms (UV/VIS Identity)

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N3C=CC(=O)NC3=O)(c4ccccc4)c5ccc(OC)cc5

InChI

1S/C39H47N4O8P/c1-27(2)43(28(3)4)52(49-24-10-22-40)51-34-25-37(42-23-21-36(44)41-38(42)45)50-35(34)26-48-39(29-11-8-7-9-12-29,30-13-17-32(46-5)18-14-30)31-15-19-33(47-6)20-16-31/h7-9,11-21,23,27-28,34-35,37H,10,24-26H2,1-6H3,(H,41,44,45)/t34-,35+,37+,52?/m0/s1

InChI key

ZGJUDTLSWZPIDW-MVFNBKNKSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service