Skip to Content
Merck
All Photos(1)

Documents

EP022363719

Eppendorf® Safe-Lock microcentrifuge tubes

size 0.5 mL, PCR clean, colorless, pkg of 500 ea

Synonym(s):

Eppendorf Microcentrifuge Tubes, Safe-Lock Microcentrifuge Tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.22

material

polypropylene

sterility

non-sterile

feature

PCR clean

packaging

pkg of 500 ea

manufacturer/tradename

Eppendorf® 022363719

technique(s)

PCR: suitable

size

0.5 mL

color

colorless

Looking for similar products? Visit Product Comparison Guide

General description

Our Safe Lock microtubes are designed to offer excellent centrifugation stability and optimized sealing properties and ensure your valuable samples remain safe and secure.

Application

Eppendorf® Safe-Lock microcentrifuge tubes are suitable for plasmid DNA recovery.

Features and Benefits

  • The hinged safe-lock lid prevents the opening of the lid during storage and incubation
  • Polypropylene provides high resistance to chemicals, high temperatures, and mechanical stress
  • Manufactured without plasticizers, biocides, and slip agents
  • Tight lid sealing prevents evaporation during long-term storage
  • Large frosted lid and surface on the side for labeling
  • Functions between -86 °C to 100 °C
  • Autoclavable at 121°C for 20 min
  • g-Safe®: Excellent centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
Tubes snap closed and stay closed. Flat top with writing surface for positive sample identification

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service