Skip to Content
Merck
All Photos(1)

Key Documents

CLS354540

Corning® BioCoat 24 well plates

3 μm pore size, Tissue Culture (TC)-treated surface PET pkg of 24 ea (4 packs )Tissue Culture (TC)-treated surface

Synonym(s):

Cell Culture Plate, plastic coated cell culture plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.13

Product Name

Corning® BioCoat Collagen I Inserts, pore size 3 μm, PET, size 6 well

material

PET

packaging

pkg of 24 ea (4 packs )

manufacturer/tradename

Corning 354540

size

6 well

pore size

3 μm

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Application

Cell culture

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service