CLS353097
Corning® Falcon® Permeable Support
for use with 24 Well Plate, with 8.0 µm Transparent PET Membrane, sterile
Synonym(s):
Permeable Support
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
Recommended Products
sterility
sterile
feature
with 8.0 µm Transparent PET Membrane
packaging
case of 48 ea (6 packs of 8)
case of 48 ea (Individually wrapped)
manufacturer/tradename
Corning 353097
compatibility
for use with 24 Well Plate
Looking for similar products? Visit Product Comparison Guide
General description
Falcon® Permeable Support for 24 Well Plate with 8.0µm Transparent Polyester (PET) Membrane, Sterile, 8/Pack, 48/Case
Application
Cell Culture
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service