CLS353072
Corning® Falcon® Cell Culture Plate
wells, 96, Tissue Culture (TC)-treated surface, flat bottom clear wells, sterile
Synonym(s):
Cell Culture Plate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material:
flat bottom clear wells
size:
96 well
sterility:
sterile
binding type:
Tissue Culture (TC)-treated surface
feature:
wells 96
material
flat bottom clear wells
sterility
sterile
feature
wells 96
manufacturer/tradename
Corning 353072
size
96 well
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Falcon® 96 Well Cell Culture Plate, Clear, Flat Bottom, Tissue Culture Treated, Sterile
Application
Cell Culture
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service