Skip to Content
Merck
All Photos(2)

Key Documents

CLS3236

Corning® CellCube® Tubing Manifold

3/8”ID Thermoelastic Tubing, Cross Connection, Aseptic, sterile

Synonym(s):

lab equipment accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.13

material

polystyrene
tubing (3/8”ID Thermoelastic)

sterility

sterile

packaging

pkg of 4 ea

manufacturer/tradename

Corning 3236

General description

  • CellCube Cross Manifold
  • 4 Aseptic Connectors

Application

Adherent Cell Scale Up

Legal Information

CELLCUBE is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service