Skip to Content
Merck
All Photos(1)

Key Documents

CLS10031

Corning® HYPERStack cell culture vessel

surface (non-treated), sterile

Synonym(s):

cell culture bioreactors, cell culture flasks, cell culture vessels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116155
NACRES:
NB.13

material

flat cap
high-density polyethylene cap
rectangular bottom flask
straight neck

sterility

sterile

feature

cap
barcoded

packaging

case of 4 ea

manufacturer/tradename

Corning 10031

flask surface area

(1720 cm2)

flask working volume

(560 mL)

surface area

1720 cm2

binding type

non-treated surface

compatibility

designed for manual cell culture applications

Looking for similar products? Visit Product Comparison Guide

General description

The High Yield PERformance Flask Manual (HYPERFlask M) cell culture vessel is designed for manual use and is commonly used on benchtops. It provides a large surface area for cell culture and is suitable for high-density cell growth.

Application

Corning® HYPERStack cell culture vessel has been used for culturing HEK293 cells for adeno-associated viral vector (AAV) vector production.

Features and Benefits

  • Ergonomic Cap: The liner cap of the vessel features a smooth texture, allowing for more ergonomic opening and closing by the operator.
  • Adaptor Grid: The new design of the adaptor grid enables faster filling and emptying of the vessel while reducing foam generation.
  • Bar Coded: Each individual flask is barcoded for easy reading by the unaided eye or a handheld bar code reader, enabling traceability.
  • Crafted with USP Class VI Materials: The flasks are manufactured and assembled in a Certified Class 100,000 clean room using materials that meet USP Class VI standards.
  • Sterile: The flasks undergo gamma irradiation to achieve a sterility assurance level (SAL) of 10-6, ensuring their sterility.
  • Clean Room Packaging: Each pack of flasks is double bagged to maintain cleanliness during transportation and storage.
  • Innovative 10-Layer Design: The flasks feature a unique 10-layer design with interconnected polystyrene growth surfaces.
  • Increased Cell Yield: The innovative design results in a 10-fold higher cell yield, significantly increasing productivity and capacity.
  • Time and Space Savings: By handling a single flask instead of 10 traditional 175 cm² flasks, processing time and incubator storage space are reduced, leading to time and space savings.

Legal Information

Corning is a registered trademark of Corning, Inc.
HYPERStack is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Florea et al.
Molecular therapy. Methods & clinical development, 28, 146-159 (2023-01-20)
The adeno-associated viral vector (AAV) provides a safe and efficient gene therapy platform with several approved products that have marked therapeutic impact for patients. However, a major bottleneck in the development and commercialization of AAV remains the efficiency, cost, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service