Skip to Content
Merck
All Photos(1)

Documents

143B

91112502, human bone, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

143B, 91112502

biological source

human bone

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human bone osteosarcoma TK-

Cell Line Description

Cells are thymidine kinase deficient (TK-) and resistant to BUdR.143B and HOS (Sigma Catalogue number 87070202) originated from the same host: 143B is TK- and originated indirectly from HOS which is TK+.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 12
D13S317: 12
D16S539: 10,13
D5S818: 13
D7S820: 11,12
THO1: 6
TPOX: 11
vWA: 18

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 15ug/ml Bromo-deoxy Uridine (BUdR) + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:8 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service