Skip to Content
Merck
All Photos(1)

Key Documents

Bu (IMR-31)

89051704, buffalo lung, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
buffalo lung
growth mode:
Adherent
karyotype:
2n = 60
morphology:
Fibroblast
products:
Not specified
receptors:
Not specified

Product Name

Bu (IMR-31), 89051704

biological source

buffalo lung

growth mode

Adherent

karyotype

2n = 60

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Buffalo lung

Cell Line Description

Derived from female Buffalo yearling. The cells have a finite life span of a minimum of 30 passages and are susceptible to a range of viruses including HSV, vaccinia, PRV, BVDV, para-influenza 3, bovine enterovirus (M62/4); also IBRV and VSV (Indiana). The cell line produces both detectable BVDV antigens and infectious BVDV.

Application

Virus replication studies

Culture Medium

McCoy′s 5a + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service