Skip to Content
Merck
All Photos(4)

Documents

AV32784

Sigma-Aldrich

Anti-ACSL1 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ACS1, Anti-Acyl-CoA synthetase long-chain family member 1, Anti-FACL1, Anti-FACL2, Anti-LACS, Anti-LACS1, Anti-LACS2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

78 kDa

species reactivity

human, mouse, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACSL1(2180)

Related Categories

General description

ACSL1 is an isozyme that belongs to the long-chain fatty-acid-coenzyme A ligase family. This enzyme converts free long-chain fatty acids into fatty acyl-CoA esters. Thus, it regulates lipid synthesis and fatty acid breakdown. Studies in bovine mammary glands have reported that ACSL1 modulates the channelling of fatty acids towards milk fat formation.
Rabbit Anti-ACSL1 (AB1) antibody recognizes pig, bovine, zebrafish, human, mouse, rat, and canine ACSL1.

Immunogen

Synthetic peptide directed towards the C terminal region of human ACSL1

Application

Rabbit Anti-ACSL1 (AB1) antibody can be used for western blot applications at a concentration of 1 μg/ml.

Biochem/physiol Actions

ACSL1 encodes an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation.The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: GSFEELCRNKDVKKAILEDMVRLGKDSGLKPFEQVKGITLHPELFSIDNG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massimo Bionaz et al.
The Journal of nutrition, 138(6), 1019-1024 (2008-05-22)
The lactating bovine mammary gland is a formidable triacylglycerol-synthesizing machine and, as such, represents an ideal model for studying putative functions of distinct isoforms of solute carrier family 27 transporters [(SLC27A) 1, 2, 3, 5, 6], long chain acyl-CoA synthetases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service