Skip to Content
Merck
All Photos(1)

Key Documents

NIST1880B

Portland cement

NIST® SRM® 1880b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 5 vials x 5 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

A unit of SRM 1880b contains five sealed vials, each containing ~ 5 grams of Portland cement finely ground to pass through a 75 µm (No. 200) sieve. For more information, pleases refer to the COA and SDS.

SRM 1880B_cert

SRM 1880B_SDS

Application

The Standard Reference Material (SRM) is primarily intended to calibrate or evaluate methods used for analyzing cements and materials with similar matrix.

Features and Benefits

  • NIST certified mass fraction values for the elements in Portland cement are metrologically traceable to the SI derived unit of mass fraction and expressed as a percentage.
  • Reference mass fraction values and information mass fraction values for various constituents present in cement are provided.
  • Maintenance and expiration of SRM certification, instructions for handling, storage, and use are provided on NIST certificate.

Other Notes

Certified for the analytes listed below.
Silicon dioxide (SiO2), Aluminium oxide (Al2O3), Ferrous oxide (Fe2O3), Calcium oxide (CaO), Magnesium oxide (MgO), Sulfur trioxide (SO3), Sodium oxide (Na2O), Potassium oxide (K2O), Titanium oxide (TiO2), Potassium pentaoxide (P2O5), Manganese oxide (Mn2O3), Chlorine (Cl), Chromium oxide (Cr2O3)

Reference/Informational value is provided for the following analytes.
Insoluble residue, Free CaO, Fluoride (F), Strontium oxide (SrO), Sulfide Sulfur (S), Zinc oxide (ZnO)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service