Skip to Content
Merck
All Photos(1)

Key Documents

937037

Sigma-Aldrich

cataCXium A Pd G4 ChemBeads

Synonym(s):

cataCXium A Pd G4, [di(adamantan-1-yl)(butyl)phosphine](methanesulfonato-κO)[2′-(methylamino)-2-biphenylyl]palladium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H54NO3PPdS
CAS Number:
Molecular Weight:
742.30
UNSPSC Code:
12352100

form

solid

Quality Level

composition

, 4-6 wt. % (loading of catalyst)

reaction suitability

reagent type: catalyst
core: palladium
reaction type: Cross Couplings

SMILES string

CCCCP([C@@]12C[C@H]3C[C@@H](C1)C[C@@H](C2)C3)[C@@]4(C5)C[C@H]6C[C@H](C[C@@H]5C6)C4.CNC7=C(C8=CC=CC=C8[Pd]OS(C)(=O)=O)C=CC=C7

InChI

1S/C24H39P.C13H12N.CH4O3S.Pd/c1-2-3-4-25(23-11-17-5-18(12-23)7-19(6-17)13-23)24-14-20-8-21(15-24)10-22(9-20)16-24;1-14-13-10-6-5-9-12(13)11-7-3-2-4-8-11;1-5(2,3)4;/h17-22H,2-16H2,1H3;2-7,9-10,14H,1H3;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

ROZFJYIWIVSDBR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

A fourth generation (G4) Buchwald precatalyst on glass beads that is similar to the third generation (G3) precatalysts except that the amino group on the biphenyl backbone is methylated. This modification helps to prevent the limitations found when using the third generation precatalysts. It is air, moisture, and thermally-stable and shows good solubility in common organic solvents.

Application

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
For general uses, product is also available in powdered form (900349)

Features and Benefits

For feature and benefits, please visit the technical article on Buchwald G4 Precatalysts. https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/cross-coupling/g3-and-g4-buchwald-precatalysts

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service