Skip to Content
Merck
All Photos(1)

Documents

34566

Supelco

Flonicamid Metabolite TFNG

PESTANAL®, analytical standard

Synonym(s):

N-[[4-(Trifluoromethyl)-3-pyridinyl]carbonyl]glycine, 4-(Trifluoromethyl)nicotinoyl glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7F3N2O3
CAS Number:
Molecular Weight:
248.16
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

O=C(NCC(O)=O)C1=C(C(F)(F)F)C=CN=C1

InChI

1S/C9H7F3N2O3/c10-9(11,12)6-1-2-13-3-5(6)8(17)14-4-7(15)16/h1-3H,4H2,(H,14,17)(H,15,16)

InChI key

AXMBYGGSBXWTEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flonicamid Metabolite TFNG is one of the metabolites of flonicamid, which is a selective aphicide.

Application

TFNG may be used as a reference standard for the determination of the analyte:
  • In vegetables using QuEChERS and reversed-phase liquid chromatography coupled to tandem mass spectrometry.
  • In dried hops by liquid chromatography coupled to tandem mass spectrometry.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of flonicamid and its metabolites in dried hops by liquid chromatography- tandem mass spectrometry.
Hengel MJ and Miller M
Journal of Agricultural and Food Chemistry, 55(20), 8033- 8039 (2007)
Simultaneous determination of flonicamid and its metabolites in vegetables using QuEChERS and reverse-phase liquid chromatography- tandem mass spectrometry.
Xu Y, et al.
Journal of Chromatography A, 1218(38), 6663- 6666 (2011)
Shanshan Wang et al.
Ecotoxicology and environmental safety, 161, 420-429 (2018-06-17)
Flonicamid, a novel selective systemic pesticide, can effectively control a broad range of insect pests. However, the dissipation behaviors and the terminal residues of flonicamid and its metabolites in some crops and soils remain unclear. Herein, an easy, sensitive and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service