Skip to Content
Merck
All Photos(1)

Key Documents

30-2290

Sigma-Aldrich

Thymolphthalein

JIS special grade

Synonym(s):

5′,5′′-Diisopropyl-2′,2′′-dimethylphenolphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H30O4
CAS Number:
Molecular Weight:
430.54
Beilstein:
359413
MDL number:
UNSPSC Code:
12171502
PubChem Substance ID:

grade

JIS special grade

form

solid

availability

available only in Japan

mp

251-253 °C (lit.)

SMILES string

CC(C)c1cc(c(C)cc1O)C2(OC(=O)c3ccccc23)c4cc(C(C)C)c(O)cc4C

InChI

1S/C28H30O4/c1-15(2)20-13-23(17(5)11-25(20)29)28(22-10-8-7-9-19(22)27(31)32-28)24-14-21(16(3)4)26(30)12-18(24)6/h7-16,29-30H,1-6H3

InChI key

LDKDGDIWEUUXSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Babaei et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 22(7), 955-959 (2006-07-14)
A novel, sensitive and selective adsorptive stripping procedure for simultaneous determination of copper, bismuth and lead is presented. The method is based on the adsorptive accumulation of thymolphthalexone (TPN) complexes of these elements onto a hanging mercury drop electrode, followed
Y Tsuda et al.
The Japanese journal of antibiotics, 46(1), 1-7 (1993-01-01)
We presented a new colorimetric bioassay of aminoglycoside antibiotics, which were represented by netilmicin (NTL) in this study, based on the discoloration of thymolphthalein (TP) in paper (indicator-disc) by carbon dioxide produced by Bacillus subtilis. To evaluate the amount of
P Balderas-Hernández et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(3), 569-577 (2004-01-30)
The detailed analysis of the experimental spectrophotometric data obtained from solutions containing the acid-base indicator thymol blue (TB) and mercury(II) (Hg(II)) coupled with data processing by means of the SQUAD program, a chemical model was determined that includes the formation
Naoki Hirayama et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 28(6), 541-543 (2012-06-26)
The use of a hydrophilic ionic liquid (IL), 1-butyl-3-methylimidazolium chloride (C(4)mimCl), as a salting-out phase separation agent to a water-tetrahydrofuran homogeneous system was studied for possible applications to novel aqueous biphasic extraction separation. The IL showed a salting-out phase-separation ability.
Shangjie Xu et al.
Macromolecular bioscience, 7(8), 968-974 (2007-08-01)
A simple general synthetic concept to build dendritic core-shell architectures with pH-labile linkers based on hyperbranched PEI cores and biocompatible PEG shells is presented. Using these dendritic core-shell architectures as nanocarriers, the encapsulation and transport of polar dyes of different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service