Skip to Content
Merck
All Photos(1)

Key Documents

05-7180

Sigma-Aldrich

Cyclohexanol

SAJ first grade, ≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11OH
CAS Number:
Molecular Weight:
100.16
Beilstein:
906744
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.5 (vs air)

vapor pressure

0.98 mmHg ( 25 °C)

Assay

≥95.0%

form

liquid

autoignition temp.

572 °F

expl. lim.

12.25 %

availability

available only in Japan

refractive index

n20/D 1.465 (lit.)

pH

6.5 (20 °C, 40 g/L)

bp

160-161 °C (lit.)

mp

20-22 °C (lit.)

density

0.948 g/mL at 25 °C (lit.)

SMILES string

OC1CCCCC1

InChI

1S/C6H12O/c7-6-4-2-1-3-5-6/h6-7H,1-5H2

InChI key

HPXRVTGHNJAIIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

147.2 °F - closed cup - DIN 51794

Flash Point(C)

64 °C - closed cup - DIN 51794

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew R Carstens et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(28), 8732-8737 (2015-07-01)
A new paradigm in oncology establishes a spectrum of tumorigenic potential across the heterogeneous phenotypes within a tumor. The cancer stem cell hypothesis postulates that a minute fraction of cells within a tumor, termed cancer stem cells (CSCs), have a
Rohan Karande et al.
Biotechnology and bioengineering, 113(1), 52-61 (2015-07-15)
The applications of biocatalysts in chemical industries are characterized by activity, selectivity, and stability. One key strategy to achieve high biocatalytic activity is the identification of novel enzymes with kinetics optimized for organic synthesis by Nature. The isolation of novel
Xiao-Kun She et al.
Journal of chromatography. A, 1423, 31-38 (2015-11-13)
Porous lead(II)-based metal-organic nanotubes (Pb-MONTs) were, for the first time, used as an adsorbent for dispersive solid-phase extraction (d-SPE) of polybrominated diphenyl ethers (PBDEs) at trace levels from environmental water samples. Gas chromatography-negative chemical ionization-mass spectrometry (GC-NCI-MS) was applied for
Aamer Saeed et al.
Journal of fluorescence, 25(4), 1045-1053 (2015-06-19)
A series of dyes using tetrachloroperylene dianhydride as fluorescent chromophore was synthesized by the substitution of suitable aliphatic and alicyclic alcohols in alkaline medium, and evaluation of dyes was done for their optical, electrochemical and thermal properties. These dyes exhibited
Martin Enmark et al.
Journal of chromatography. A, 1400, 131-139 (2015-05-24)
In SFC the sample cannot be dissolved in the mobile phase, so it is often dissolved in pure modifier, or another liquid, sometimes resulting in serious distortions of the eluted peak profiles already at moderately high injection volumes. It is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service