Skip to Content
Merck
All Photos(1)

Key Documents

MWTS

Millipore

mPAGE® Mini Wet Transfer System

for transfer of proteins from a mini acrylamide gel to blotting membrane, accommodates two wet transfer cassettes

Synonym(s):

transfer device, wet transfer system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105311
NACRES:
NA.32
Pricing and availability is not currently available.

AC/DC input

100 watts (maximum power limit)
300 V (maximum voltage limit)

packaging

pkg of 1 system
pkg of 1 ea ( for transferring up to 2 gels)

manufacturer/tradename

Millipore

parameter

70 °C ambient temp. limit

technique(s)

immunoblotting: suitable
western blot: suitable

L × W × H

16 cm × 12 cm × 18 cm

buffer volume

1255 mL , Wet transfer module with freezer pack
1405 mL , Wet transfer module without freezer pack

tank L × W × H

17 cm × 13 cm × 15.5 cm

General description

The mPAGE® Mini Wet Transfer System accommodates two gel holder cassettes for electrophoretic transfer of proteins from mini gels to PVDF or nitrocellulose blotting membrane using mPAGE® Transfer Buffer or a compatible transfer buffer.

Components

The system includes the following components:
  • mPAGE® Tank Lid with Electrode Cables (1)
  • mPAGE® Replacement Tank (1)
  • mPAGE® Blot Cassette (1)
  • mPAGE® Blot Cassette Clips (2)
  • mPAGE® Macroporous Sponge (5)
  • mPAGE® Freezer Pack (2)

Compatibility

mPAGE Mini Gel Tank

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service