Skip to Content
Merck
All Photos(1)

Key Documents

MSEHNFX40

Millipore

Multiscreen® 96 well Plate, solid bottom

flat bottom clear polystyrene plate, non-sterile, pkg of 40 plates, high binding surface

Synonym(s):

MultiScreen Solid Plate, Solid Bottom 96-Well Plate, Solid Bottom Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NB.15
material:
flat bottom clear polystyrene plate
plate size:
96 wells
sterility:
non-sterile
binding type:
high binding surface
Pricing and availability is not currently available.

material

flat bottom clear polystyrene plate

Quality Level

sterility

non-sterile

product line

MultiScreen®

packaging

pkg of 40 plates

technique(s)

ELISA: suitable

plate size

96 wells

binding type

high binding surface

General description

MultiScreen® 96-well ELISA microplates meet industry standard dimensions for use in common biochemical assay platforms, including automated liquid handling, microplate readers, and other microplate compatible devices. ELISA microplates are available with medium or high binding capabilities.

Physical form

Color Code:Clear

Other Notes

Automation Compatible:Yes

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service