Skip to Content
Merck
All Photos(1)

Key Documents

MP0DPIL0T

Millipore

Pilot Scale Holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690
Pricing and availability is not currently available.

material

304 stainless steel

Quality Level

fitting

1.5 in. gauge port sanitary flange
1.5 in. inlet/outlet sanitary flange

compatibility

for use with Pod Filters

Application

For Pod configurations up to 2.2 m2

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service