KMEFHAW10B
EZ-Fit® Manifold Kit
Contains 4 boxes of EZ-Fit™ 100 mL Funnels with bulk packaging, EZ-Fit® 3-head Manifold with filtration heads
Synonym(s):
EZ-Fit® Filtration system for bioburden testing, EZ-Fit® Manifold Starting Kit, Filtration Kit, Filtration Unit
About This Item
Recommended Products
Agency
ISO 7004
sterility
electron beam irradiated
packaging
pkg of 4 units (bulk packaging)
manufacturer/tradename
EZ-Fit®
volume
100 mL
pore size
0.45 μm pore size
application(s)
bioburden testing
food and beverages
pharmaceutical
water monitoring
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
The EZ-Fit® Filtration device is designed to fit onto EZ-Fit® Manifold, which features specific EZ-Fit® Filtration Heads.
The state-of-the-art design is optimized to secure laboratory workflow, avoid cross contamination and save testing time thereby producing reliable microbiological results.
After the filtration process is completed, the membrane can be easily transferred onto an agar plate for microbial growth.
Application
Packaging
EZFITBASE3 x1
EZFITMVHE3 x1
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service